Skip to Content
Merck
All Photos(1)

Key Documents

203392

Sigma-Aldrich

(+)-Blebbistatin

The inactive enantiomer of (±)-Blebbistatin.

Synonym(s):

(+)-Blebbistatin

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€221.00

€221.00


Estimated to ship on29 May 2025



Select a Size

Change View
1 MG
€221.00

About This Item

Empirical Formula (Hill Notation):
C18H16N2O2
CAS Number:
Molecular Weight:
292.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€221.00


Estimated to ship on29 May 2025


Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

methanol: 1.5 mg/mL
100% DMSO: 100 mg/mL
90% DMSO: 75 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

N2(CC[C@]3(C2=Nc4c(cc(cc4)C)C3=O)O)c1ccccc1

InChI

1S/C18H16N2O2/c1-12-7-8-15-14(11-12)16(21)18(22)9-10-20(17(18)19-15)13-5-3-2-4-6-13/h2-8,11,22H,9-10H2,1H3/t18-/m0/s1

InChI key

LZAXPYOBKSJSEX-SFHVURJKSA-N

General description

The inactive enantiomer of (±)-Blebbistatin (Cat. No. 203390). Useful as a negative control for the active enantiomer (Cat. No. 203391).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Inactive enantiomer of (±)-Blebbistatin
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Reconstitution

Following reconstitution in methanol, aliquot and freeze (-20°C). Stock solutions in 90% DMSO or methanol are stable for up to 1 month at -20°C. Stock solutions in 100% DMSO are unstable; reconstitute just prior to use.

Other Notes

Shu, S., et al. 2005. Proc. Natl. Acad. Sci. USA102, 1472.
Jacobelli, J., et al. 2004. Nat. Immunol.5, 531.
Kovacs, M., et al. 2004. J. Biol. Chem.279, 35557.
Straight, A.F., et al. 2003. Science299, 1743.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Liu et al.
Kidney diseases (Basel, Switzerland), 6(6), 422-433 (2020-12-15)
Levels of urinary microvesicles, which are increased during various kidney injuries, have diagnostic potential for renal diseases. However, the significance of urinary microvesicles as a renal disease indicator is dampened by the difficulty to ascertain their cell source. The aim

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service