Skip to Content
Merck
All Photos(1)

Documents

203188

Sigma-Aldrich

Biotin-X-NHS

Synonym(s):

Biotin-X-NHS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30N4O6S
CAS Number:
Molecular Weight:
454.54
UNSPSC Code:
41116133

Assay

≥90% (NMR)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

This product has been discontinued.



Used to biotinylate amino acids, peptides, or proteins by reacting with primary amines under mild conditions. Inserts a 6-atom spacer between biotin and the target ligand, thereby alleviating steric hindrance. Useful for red blood cell survival studies.

Analog of Biotin-NHS ester containing a 6-aminocaproic acid "X" spacer arm. Used to biotinylate amino acids, peptides, or proteins by reacting with primary amines under mild conditions. Inserts a six atom spacer between biotin and the target ligand that helps alleviate steric hindrance.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Unstable in solution; reconstitute just prior to use.

Other Notes

Hoffmann-Fezer, G., et al. 1997. Ann. Hematol.74, 231.
Fabris, F., et al. 1992. Thromb. Res. 66, 409.
Smith, J.S., et al. 1991. Anal. Biochem.197, 247.
Hastie, A.T., et al. 1990. Cell Motility Cytoskel.17, 317.
Ghebrehiwet, B., et al. 1988. J. Immunol. Methods110, 251.
Liu, Y.S.V. and Green, A. 1985. Clin. Chem.31, 202.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service