Skip to Content
Merck
All Photos(2)

Key Documents

200488

Sigma-Aldrich

Bestatin Hydrochloride

InSolution, ≥98%, a metalloprotease inhibitor

Synonym(s):

InSolutionBestatin, Hydrochloride, [(2S,3R)-3-Amino-2-hydroxy-4-phenylbutanoyl]-Leu, HCl

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€225.00

€225.00


Estimated to ship on23 May 2025



Select a Size

Change View
10 MG
€225.00

About This Item

Empirical Formula (Hill Notation):
C16H24N2O4 · xHCl
Molecular Weight:
308.37 (free base basis)
UNSPSC Code:
12352202
NACRES:
NA.77

€225.00


Estimated to ship on23 May 2025


Quality Level

Assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
protect from light

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC(C)C[C@@H](C(O)=O)NC([C@@H](O)[C@H](N)CC1=CC=CC=C1)=O.[xHCl]

General description

A metalloprotease inhibitor. Reversibly binds to cell surfaces and competitively inhibits surface aminopeptidases, notably aminopeptidase B and leucine aminopeptidase. Also acts as a potent aminopeptidase N inhibitor. Activates macrophages and T lymphocytes. Has anti-tumor properties. Effective at a concentration of 1-10 µM. The solid form of this compound (Cat. No. 200484) is also available.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Metalloproteases
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied as a 50 mM (10 mg/580 µL) solution of Bestatatin, HCl in H₂O.

Reconstitution

Following initial, aliquot and freeze (-20°C). Aliquots are stable for up to 6 months at -20°C.

Other Notes

Merck Index: 14, 9842.
Inoue, T., et al. 1994. J. Clin. Endocrinol. Metab.79, 171.
Taylor, A., et al. 1993. Biochemistry32, 784.
Ishida, J.I., et al. 1984. J. Chromatogr.305, 381.
Umezawa, H. 1982. Annu. Rev. Microbiol.36, 75.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service