Skip to Content
Merck
All Photos(1)

Key Documents

197225

Sigma-Aldrich

Bcl-2 Binding Peptide, Cell-Permeable, Negative Control

A cell-permeable, mutated cpm-1285 peptide analog (Leu¹⁵¹ replaced by Ala) that does not induce apoptosis and displays nearly 15-fold reduction in Bcl-2 binding.

Synonym(s):

Bcl-2 Binding Peptide, Cell-Permeable, Negative Control, Decanoyl-KN¹⁴⁰LWAAQRYGREARRMSDEFEGSFKGL¹⁶⁵-OH, cpm-1285m, cpm-1285m, Decanoyl-KN¹⁴⁰LWAAQRYGREARRMSDEFEGSFKGL¹⁶⁵-OH

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€215.00

€215.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€215.00

About This Item

Empirical Formula (Hill Notation):
C150H234N44O42S
Molecular Weight:
3357.80
UNSPSC Code:
12352200
NACRES:
NA.77

€215.00


Estimated to ship on24 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

off-white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable, mutated cpm-1285 peptide analog (Leu151 replaced by Ala) that does not induce apoptosis and displays nearly 15-fold reduction in Bcl-2 binding. Suitable for use as a negative control for Cat. No. 197220.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
A negative control for Cat. No. 197220 (Bcl-2 Binding Peptide, Cell-Permeable).
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

CH₃(CH₂)₈CONH-Lys-Asn-Leu-Trp-Ala-Ala-Gln-Arg-Tyr-Gly-Arg-Glu-Ala-Arg-Arg-Met-Ser-Asp-Glu-Phe-Glu-Gly-Ser-Phe-Lys-Gly-Leu-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot, flush with an inert gas, and freeze at -20°C. Stock solutions are stable for up to 2 weeks at -20°C.

Other Notes

Wang, J-L., et al. 2000. Cancer Res.60, 1498.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service