Skip to Content
Merck
All Photos(1)

Key Documents

196876

Sigma-Aldrich

BAY 41-2272

A cell-permeable pyrazolopyridinylpyrimidine compound that acts as a selective and potent stimulator of soluble guanylate cyclase (effective dose ~ 0.1 nM to 100 µM using recombinant soluble guanylate cyclase).

Synonym(s):

BAY 41-2272, 5-Cyclopropyl-2-[1-(2-fluorobenzyl)-1H-pyrazolo[3,4-b]pyridin-3-yl]pyrimidin-4-ylamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€253.00

€253.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€253.00

About This Item

Empirical Formula (Hill Notation):
C20H17FN6
CAS Number:
Molecular Weight:
360.39
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

€253.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Fc1c(cccc1)C[n]2nc(c5c2nccc5)c3nc(c(cn3)C4CC4)N

InChI

1S/C20H17FN6/c21-16-6-2-1-4-13(16)11-27-20-14(5-3-9-23-20)17(26-27)19-24-10-15(12-7-8-12)18(22)25-19/h1-6,9-10,12H,7-8,11H2,(H2,22,24,25)

InChI key

ATOAHNRJAXSBOR-UHFFFAOYSA-N

General description

A cell-permeable pyrazolopyridinylpyrimidine compound that acts as a selective and potent stimulator of soluble guanylate cyclase (effective dose ~ 0.1 nM to 100 µM using recombinant soluble guanylate cyclase). The mode of activation is NO-independent and appears to be mediated through direct binding to the α1 and α2 subunits. Shown to be effective in treating numerous cardiovascular conditions in animal models. Inhibits phenylephrine-induced constriction of rabbit aortic rings (IC50 = 304 nM) and blocks collagen-induced aggregation of human platelets (IC50 = 36 nM). Does not inhibit phosphodiesterases.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Soluble guanylate cyclase
Product does not compete with ATP.
Reversible: no
Target IC50: 304 nM inhibiting phenylephrine-induced constriction of rabbit aortic rings; 36 nM in blocking collagen-induced aggregation of human platelets

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Boerrigter, G., et al. 2003. Circulation107, 686.
Kalsi, J.S., et al. 2003. J Urol.169, 761.
Koglin, M., et al. 2002. Biochem. Biophys. Res. Commun.292, 1057.
Becker, E.M., et al. 2001. BMC Pharmacol.1, 13.
Stasch, J.P., et al. 2001. Nature410, 212.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service