Skip to Content
Merck
All Photos(1)

Key Documents

196811

Sigma-Aldrich

Bax-Inhibiting Peptide, Negative Control

A cell-permeable negative control of the Bax-Inhibiting Peptide, V5. Does not suppress Bax-mediated apoptosis (~200 µM).

Synonym(s):

Bax-Inhibiting Peptide, Negative Control, BIP-NC, H-IPMIK-OH

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€121.00

€121.00


Estimated to ship on21 May 2025



Select a Size

Change View
5 MG
€121.00

About This Item

Empirical Formula (Hill Notation):
C28H52N6O6S
Molecular Weight:
600.81
UNSPSC Code:
12352202
NACRES:
NA.77

€121.00


Estimated to ship on21 May 2025


Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 1 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable negative control of the Bax-Inhibiting Peptide, V5 (Cat. No. 196810). Does not suppress Bax-mediated apoptosis (~200 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Negative control of the Bax-Inhibiting Peptide, V5 (Cat. No. 196810)
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H-Ile-Pro-Met-Ile-Lys-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Sawada, M., et al. 2003. Nature Cell Biol.5, 352.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service