Skip to Content
Merck
All Photos(2)

Key Documents

19-142

Sigma-Aldrich

LY 294002

Synonym(s):

NSC 697286, PI3K inhibitor, SF 1101

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41
Pricing and availability is not currently available.

form

solid

Quality Level

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

shipped in

wet ice

Application

Specific and potent inhibitor of PI 3-kinase.

Biochem/physiol Actions

Inhibitor Type: Kinase

Physical form

C19H17NO3

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

487.4 °F

Flash Point(C)

253.0 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen Jiang et al.
Bioengineered, 12(2), 10264-10284 (2021-11-02)
Chronic skin ulcers are a primary global health problem. Velvet antler polypeptide (VAP) regulates endothelial cell migration and angiogenic sprout. Adipose-derived stem cells (ADSCs) are reported to make pivotal impacts upon wound healing. This study aimed to explore the role
Jie Ni et al.
Molecular medicine reports, 22(2), 1277-1284 (2020-07-07)
MicroRNAs (miRNAs/miRs) are small noncoding RNAs 19‑24 nucleotides in length, which can play an important role in tumor development. However, the influence of miRNAs on the tumorigenicity of ovarian cancer cells has not been fully elucidated. Previously, DEAD box protein (DDX)
Hongxiu Han et al.
The American journal of pathology, 191(3), 515-526 (2020-12-22)
Nerve infiltration into the tumor is a common feature of the tumor microenvironment. The mechanisms of axonogenesis in breast cancer remain unclear. We hypothesized that vascular endothelial growth factor (VEGF), as well as nerve growth factor (NGF), is involved in
Shang Li et al.
NPJ biofilms and microbiomes, 9(1), 47-47 (2023-07-09)
Knowledge gaps that limit the development of therapies for polycystic ovary syndrome (PCOS) concern various environmental factors that impact clinical characteristics. Circadian dysrhythmia contributes to glycometabolic and reproductive hallmarks of PCOS. Here, we illustrated the amelioration of Limosilactobacillus reuteri (L.
Qize Wei et al.
Molecular biology of the cell, 13(2), 683-697 (2002-02-21)
We ectopically expressed the transcription factor Pitx2a, one of the Pitx2 isoforms, in HeLa cells by using a tetracycline-inducible expression system and examined whether Pitx2a was capable of modulating Rho GTPase signaling and altering the cell's cytoskeleton. Ectopic expression of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service