Skip to Content
Merck
All Photos(1)

Key Documents

189497

Sigma-Aldrich

Autophagy Inducer, STF-62247

A cell-permeable autophagy inducer that selectively induces non-apoptotic cell death in VHL-deficient, but not VHL-expressing, renal carcinoma cells both in vitro and in mice in vivo.

Synonym(s):

Autophagy Inducer, STF-62247, N-(3-Methylphenyl)-4-(4-pyridinyl)-2-thiazolamine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€122.00

€122.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€122.00

About This Item

Empirical Formula (Hill Notation):
C15H13N3S
CAS Number:
Molecular Weight:
267.35
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€122.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

beige

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[s]1c(nc(c1)c3ccncc3)Nc2cc(ccc2)C

InChI

1S/C15H13N3S/c1-11-3-2-4-13(9-11)17-15-18-14(10-19-15)12-5-7-16-8-6-12/h2-10H,1H3,(H,17,18)

InChI key

KATNUHQNJGNLPW-UHFFFAOYSA-N

General description

A cell-permeable autophagy inducer that selectively induces non-apoptotic cell death in VHL- (von Hippel-Lindau) deficient, but not VHL-expressing, renal carcinoma cells both in vitro (IC50 = 0.625 and 16 µM in RCC4 and RCC4/VHL killing, respectively) and in mice in vivo (>90% 786-O tumor growth suppression; 8 mg/kg via daily i.p.). Although STF-62247 induces autophagy regardless of cellular VHL-expression level, STF-62247 treatment is shown to result in more enhanced/acidified autolysomes formation in VHL-deficient cells, implicating an important role of VHL in maintaining normal vesicular trafficking between ER, TNG, and lysosomes.

Packaging

Packaged under inert gas

Warning

Toxicity: Toxic (F)

Other Notes

Turcotte, S., et al. 2008. Cancer Cell14, 90.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service