Skip to Content
Merck
All Photos(2)

Key Documents

189299

Sigma-Aldrich

ATR/CDK Inhibitor, NU6027

The ATR/CDK Inhibitor, NU6027, also referenced under CAS 220036-08-8, controls the biological activity of ATR/CDK. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

ATR/CDK Inhibitor, NU6027, ATR Inhibitor, NU6027, 2,6-diamino-4-cyclohexyl-methyloxy-5-nitroso-pyrimidine, CDK2 Inhibitor, NU6027

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€174.00

€174.00


Estimated to ship on18 May 2025



Select a Size

Change View
10 MG
€174.00

About This Item

Empirical Formula (Hill Notation):
C11H17N5O2
CAS Number:
Molecular Weight:
251.28
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€174.00


Estimated to ship on18 May 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

red-purple

solubility

DMSO: 10 mg/mL, clear, blue

shipped in

ambient

storage temp.

−20°C

SMILES string

Nc1nc(c(c(n1)OCC2CCCCC2)N=O)N

InChI

1S/C11H17N5O2/c12-9-8(16-17)10(15-11(13)14-9)18-6-7-4-2-1-3-5-7/h7H,1-6H2,(H4,12,13,14,15)

InChI key

DGWXOLHKVGDQLN-UHFFFAOYSA-N

General description

A cell-permeable, pyrimidine derivative that acts as an ATP-competitive CDK Inhibitor (Ki = 2.5 µM for CDK1, and Ki = 1.3 µM for CDK2), and demonstrates cytostatic properties among a panel of 57 cancer cell lines. Also inhibits ATR kinase (IC50 = 6.7 µM in MCF7 cells), but does not interfere with irradiation-induced autophosphorylation of DNA-PK or ATM. Potentiates a range of DNA-damaging cytotoxic drugs such as hydroxyurea (1.8-fold) and cisplatin (1.4-fold) at 10 µM, but not the anti-mitotic paclitaxel. Attenuates G2/M arrest following DNA damage and inhibits RAD51 focus formation. Shown to be synthetically lethal when DNA single-strand break repair is impaired in PARP-inhibited cells.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Peasland, A., et al. 2011., Br J Cancer. 105, 372.
Arris, C.E, et al. 2000., J Med Chem.43, 2797.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service