Skip to Content
Merck
All Photos(2)

Key Documents

1.88008

Supelco

CombiSolvent

methanol-free solvent for volumetric Karl Fischer titration with one component reagents Aquastar®

Synonym(s):

Solvent Combination

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115601
NACRES:
NA.21

Quality Level

product line

Aquastar®

form

liquid

reaction suitability

reaction type: volumetric

technique(s)

Karl-Fischer titration: suitable
titration: suitable

pH

5.8-6.3 (20 °C in H2O)

transition temp

flash point 12 °C

density

0.80 g/cm3 at 20 °C

format

single component solution

storage temp.

15-25°C

Legal Information

AQUASTAR is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Summary application report for analysis of moisture in Formic acid

Summary application report for analysis of moisture in 1-Methyl-2-pyrrolidone.

Summary application report for analysis of moisture in Sodium methylate

Summary application report for analysis of moisture in Sodium tert-butoxide.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service