Skip to Content
Merck
All Photos(1)

Key Documents

178493

Sigma-Aldrich

Apoptosis Activator III, Embelin

The Apoptosis Activator III, Embelin, also referenced under CAS 550-24-3, controls the biological activity of Apoptosis. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Apoptosis Activator III, Embelin, Embelin, Embelia ribes, XIAP inhibitor, Embelin, 2,5-Dihydroxy-3-undecyl-1,4-benzoquinone

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€138.00

€138.00


Estimated to ship on21 April 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€138.00

About This Item

Empirical Formula (Hill Notation):
C17H26O4
CAS Number:
Molecular Weight:
294.39
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€138.00


Estimated to ship on21 April 2025


Request a Bulk Order

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 10 mg/mL
methanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C17H26O4/c1-2-3-4-5-6-7-8-9-10-11-13-16(20)14(18)12-15(19)17(13)21/h12,18,21H,2-11H2,1H3

InChI key

IRSFLDGTOHBADP-UHFFFAOYSA-N

General description

A cell-permeable benzoquinone compound that exhibits antitumor properties. Specifically antagonizes XIAP (X-linked inhibitor of apoptosis)-mediated inhibition of caspase-9 activation by directly targeting the Smac and caspase-9 binding domain BIR3 (IC50 = 4.1 µM in a competitive binding assay with Smac peptide). Selectively induces caspase-9-mediated apoptosis in prostate cancer cells with high levels of XIAP (IC50 = 3.7 and 5.7 µM for PC-3 and LnCap, respectively), while exhibiting much less effect towards cells with low level XIAP (IC50 = 19.3 and 20.1 µM for WI-38 and PrEC, respectively). Also shown to sensitize XIAP overexpressing Jurkat cells to Etoposide -induced apoptosis. A non-competitive PCAF (p300/CBP associated factor)-specific lysine acetyltransferase (KAT) inhibitor (IC50 = 7.2 µM).
A cell-permeable, benzoquinone compound that exhibits anti-tumor properties. Shown to specifically antagonize X-linked inhibitor of apoptosis- (XIAP) mediated inhibition of caspase-9 activation by directly targeting the Smac and caspase-9 binding domain BIR3 (IC50 = 4.1 µM in a competitive binding assay using Smac peptide). Also selectively induces caspase-9-mediated apoptosis in prostate cancer cells expressing high levels of XIAP (IC50 = 3.7 µM for PC-3 cells and 5.7 µM for LnCap cells), while exhibiting lesser effects toward cells with low levels of XIAP (IC50 = 19.3 µM for WI-38 cells and 20.1 µM for PrEC cells). Reported to sensitize XIAP-overexpressing Jurkat cells to Etoposide- (Cat. No. 341245) induced apoptosis. A non-competitive PCAF (p300/CBP associated factor)-specific lysine acetyltransferase (KAT) inhibitor (IC50 = 7.2 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
BIR3
Product does not compete with ATP.
Reversible: no
Target IC50: 4.1 µM in antagonizing XIAP (X-linked inhibitor of apoptosis)-mediated inhibition of caspase-9 activation by directly targeting the Smac and caspase-9 binding domain BIR3 in a competitive binding assay with Smac peptide

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Analysis Note

Single spot by TLC

Other Notes

Modak, R., et al. 2013. ACS Chem. Biol.8, In press.
Nikolovska-Coleska, Z., et al. 2004. J. Med. Chem.47, 2430.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service