Skip to Content
Merck
All Photos(1)

Key Documents

176708

Sigma-Aldrich

Angiostatin K1-3, Human, Recombinant, E. coli

Angiostatin K1-3 is a recombinant form expressed in E. coli as a single, non-glycosylated polypeptide chain of 259 amino acids. A specific inhibitor of endothelial cell growth and angiogenesis.

Synonym(s):

Ang K1-3, K1-3, rHuAng K1-3

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
€475.00

€475.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
250 μG
€475.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.77

€475.00


Estimated to ship on28 May 2025


Request a Bulk Order

recombinant

expressed in E. coli

Quality Level

Assay

≥95% (HPLC and SDS-PAGE)

form

lyophilized powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

solubility

aqueous buffer containing 0.1% BSA: soluble
water: soluble

shipped in

wet ice

storage temp.

−20°C

General description

Recombinant, human angiostatin expressed in E. coli as a single, nonglycosylated polypeptide chain containing 259 amino acids. Represents a proteolytic fragment containing the first three kringle structures of plasminogen. Acts as a specific inhibitor of endothelial cell growth and angiogenesis. Displays increased inhibitory activity (ED50=70 nM) relative to kringles 1-4 (ED50=135 nm).

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 20 mM Sodium Acetate, 4% Mannitol, pH 5.5.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Cao, R., et al. 1999. Proc. Natl. Acad. Sci. USA96, 5728.
Cao, E., et al. 1997. J. Biol. Chem.272, 22924.
Sim, B.K., et al. 1997. Cancer Res.57, 1329.
Wu, Z., et al. 1997. Biochem. Biophys. Res. Commun.236, 651.
O′Rieilly, M.S., et al. 1994. Cell79, 315.
Peterson, T.E., et al. 1990. J. Biol. Chem.265, 6104.
Forsgren, M., et al. 1987. FEBS Lett.213, 254.
Malinowski, D.P., et al. 1984. Biochemistry23, 4243.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service