Skip to Content
Merck

Skip To

17-643

ChIPAb+ Monomethyl-Histone H3 (Lys27) - ChIP Validated Antibody and Primer Set

serum, from rabbit

Synonym(s):

H3K27me1, Histone H3 (mono methyl K27)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.52
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
human, mouse
Application:
ChIP, EMSA, FACS, IF, IHC (p), IP, WB
Citations:
8
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

antibody form

serum

clone

polyclonal

species reactivity

human, mouse

species reactivity (predicted by homology)

mammals

manufacturer/tradename

ChIPAb+, Upstate®

technique(s)

ChIP: suitable, electrophoretic mobility shift assay: suitable, flow cytometry: suitable, immunofluorescence: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... H3F3B(3021)

General description

17 kDa
All ChIPAb+ antibodies are individually validated for chromatin precipitation, every lot, every time. Each ChIPAb+ antibody set includes control primers (tested every lot by qPCR) to biologically validate your IP results in a locus-specific context. The qPCR protocol and primer sequences are provided, allowing researchers to validate ChIP protocols when using our antibody in their chromatin context. Each set also includes a negative control antibody to ensure specificity of the ChIP reaction.
The ChIPAb+ Monomethyl-Histone H3 (Lys27) set includes the anti-monomethyl-histone H3 (Lys27) antibody, a negative control antibody (normal rabbit serum), and qPCR primers which amplify a 110 base pair region within the promoter of the human β-globin gene. The monomethyl-histone H3 (Lys27) and negative control antibodies are supplied in a scalable "per ChIP" reaction size and can be used to functionally validate the precipitation of monomethyl-histone H3 (Lys27)-associated chromatin.

Immunogen

The monomethyl-histone H3 (Lys27) antiserum is made against a synthetic 2X-branched peptide containing the sequence…ARmeKSA…in which meK corresponds to monomethyl-lysine 27 of human histone H3.

Application

Chromatin Immunoprecipitation:
Successful immunoprecipitation of monomethyl-histone H3 (Lys27) associated DNA fragments was verified by qPCR using control ChIP Primers flanking the human GAPDH promoter or primers amplifying the promoter of human β-globin, which is transcriptionally inactive in HeLa cells (Please see figures). Percent Input relative to standard curves for each qPCR primer set are shown, with immunoprecipitated DNA from control serum shown as (-) and monomethyl- histone H3 (Lys27) as (+).
Please refer to the EZ-Magna A ChIP (Cat. #17-408) or EZ-ChIP (Cat. #17-371) protocol for experimental details.

Western blot analysis and peptide inhibition:
HeLa Acid extract was resolved by electrophoresis, transferred to nitrocellulose and probed with Anti-Monomethyl-Histone H3 (Lys27) (1:5000, Lane 1) or preincubated with 0.4 μM Histone H3 peptide with following modifications:
Lane 2: Linear non-modified
Lane 3: Branched monomethyl
Lane 4: Linear monomethyl
Lane 5: Branched dimethyl
Lane 6: Linear dimethyl
Lane 7: Branched trimethyl
Lane 8: Linear trimethyl
Proteins were visualized using a goat anti-rabbit secondary antibody conjugated to HRP and a chemiluminescence detection system (Please see figures).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
This ChIPAb+ Monomethyl-Histone H3 (Lys27) -ChIP Validated Antibody & Primer Set conveniently includes the antibody & the specific control PCR primers.

Biochem/physiol Actions

Monomethyl-Histone H3 (Lys27)

Packaging

25 assays per kit, ~4μL per chromatin immunoprecipitation

Physical form

Anti-monomethyl-Histone H3 (Lys27) (rabbit polyclonal IgG). One vial containing 100 μL of antiserum containing 0.05% sodium azide.

Normal Rabbit Serum. One vial containing 100 uL antiserum containing 0.05% sodium azide.

ChIP Primers, β-Globin. One vial containing 75 μL of 5 μM of each primer specific for for human β-globin promoter.
FOR: AGG ACA GGT ACG GCT GTC ATC
REV: TTT ATG CCC AGC CCT GGC TC

Preparation Note

Stable for 1 year at -20°C from date of receipt

Analysis Note

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from untreated HeLa cells (1 X 106 cell equivalents) was subjected to chromatin immunoprecipitation using 4 μL of either a normal rabbit antiserum or Anti-Monomethyl-Histone H3 (Lys27) serum and the Magna ChIP A Kit (Cat. #17-610).
Successful immunoprecipitation of monomethyl-histone H3 (Lys27) associated DNA fragments was verified by qPCR using control ChIP Primers flanking the human β-globin promoter (Please see figures).
Please refer to the EZ-Magna A ChIP (Cat. #17-408) or EZ-ChIP (Cat. #17-371).
Control
Included negative control antibody normal rabbit serum and control primers specific for human β-globin promoter.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
17-68017-68317-622
clone

polyclonal

clone

CMA306, monoclonal

clone

CMA309, monoclonal

clone

polyclonal

antibody form

serum

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

-

biological source

rabbit

biological source

mouse

biological source

mouse

biological source

rabbit

species reactivity

human, mouse

species reactivity

vertebrates, human

species reactivity

vertebrates, human

species reactivity

human, mouse

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

Gene Information

human ... H3F3B(3021)

Gene Information

-

Gene Information

-

Gene Information

human ... H3F3B(3021)


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters