Skip to Content
Merck
All Photos(1)

Documents

17-327

Sigma-Aldrich

H2A.X Phosphorylation Assay Kit (Chemiluminescence Detection)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.54

packaging

kit of 192 assay(s)

Quality Level

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable

detection method

chemiluminescent

General description

Phosphorylation of the histone variant H2A.X is a rapid and sensitive response to double strand DNA breaks. The H2A.X Phosphorylation Assay Kit (Chemiluminescence Detection) is a cell based assay formatted for microplate-based detection of levels of phosphorylated histone H2A.X.

Application

The H2A.X Phosphorylation, Chemiluminescence Detection Assay Kit is a cell-based ELISA formatted for chemiluminescent detection of relative levels of phosphorylated H2A.X in microplate cells cultures.

Components

  • Anti-phospho-Histone H2A.X (Ser139), clone JBW301 (Cat.# 05-636)
  • TBS, 20X (Cat.# 20-190)
  • 20% Tween-20 (v/v)
  • 10% BSA in TBS (Blocking Buffer)
  • LumiGLO Substrate Reagent A
  • LumiGLO Substrate Reagent B
  • Goat Anti-Mouse IgG, HRP conjugate

Quality

The H2A.X Phosphorylation, Chemiluminescence Detection Assay Kit is a cell-based ELISA formatted for chemiluminescent detection of relative levels of phosphorylated H2A.X in microplate cells cultures. Adherent cells are cultured in microplates, treated with agents that induce DNA damage/H2A.X phosphorylation, and are then fixed and permeabilized. Histone H2A.X phosphorylated at serine 139 is detected by Anti-phospho-H2A.X (Ser139), clone JBW301 and an anti-mouse-HRP (horseradish peroxidase) conjugate. The chemiluminescent HRP substrate LumiGLO is then added, and signal is measured in a microplate luminometer

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service