Skip to Content
Merck
All Photos(1)

Key Documents

1.72572

SAFC

Magnesium sulfate heptahydrate

extra pure, for tablewater, FCC

Synonym(s):

bitter salt, epsom salt, sulfuric acid magnesium salt heptahydrate, Epsom salts

Sign Into View Organizational & Contract Pricing

Select a Size

25 KG
€100.00

€100.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 KG
€100.00

About This Item

Linear Formula:
MgSO4 · 7H2O
CAS Number:
Molecular Weight:
246.47
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-298-2

€100.00


Check Cart for Availability

Request a Bulk Order

reg. compliance

FCC

Quality Level

vapor density

<0.01 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

form

solid

pH

5.0-8.0 (25 °C, 50 g/L in H2O)

solubility

710 g/L

density

1.68 g/cm3 at 20 °C

application(s)

pharma/biopharma processes

storage temp.

2-30°C

SMILES string

O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

InChI

1S/Mg.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

InChI key

WRUGWIBCXHJTDG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

We offer a range of high quality critical raw materials plus the documentation and expertise to help our customers meet stringent regulatory requirements. Our SAFC® portfolio of high-quality products withstands strict quality control procedures.

Application

Magnesium sulfate heptahydrate with a low bromide level can be used to remineralize desalinated sea water to produce table water.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service