Skip to Content
Merck
All Photos(5)

Key Documents

17-10259

Sigma-Aldrich

ChIPAb+ Acetyl-Histone H3 (Lys56) - ChIP Validated Antibody and Primer Set

from rabbit, purified by affinity chromatography

Synonym(s):

Histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/k, Histone H3/l

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.52
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

ChIPAb+
Upstate®

technique(s)

ChIP: suitable
immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... HIST1H3F(8968)

General description

All ChIPAb+ antibodies are individually validated for chromatin precipitation, every lot, every time. Each ChIPAb+ antibody set includes control primers (tested every lot by qPCR) to biologically validate your IP results in a locus-specific context. The qPCR protocol and primer sequences are provided, allowing researchers to validate ChIP protocols when using our antibody in their chromatin context. Each set also includes a negative control antibody to ensure specificity of the ChIP reaction.
The ChIPAb+ Acetyl-Histone H3 (Lys56) set includes the Acetyl-Histone H3 (Lys56) antibody, a Normal Rabbit IgG, and control primers which amplify a 152 bp region of the human cyclin B1 promoter. The Acetyl-Histone H3 (Lys56) and negative controls are supplied in a scalable "per ChIP" reaction size and can be used to functionally validate the precipitation of Acetyl-Histone H3 (Lys56) associated chromatin.

Application

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from HeLa cells (1x104, 1x105 and 1x106 cell equivalents per IP) were subjected to chromatin immunoprecipitation using 1 µL of either Normal Rabbit IgG (Part No. CS213233), or 1 µL Anti-Acetyl-Histone H3 (Lys56) (Part No.CS207337) and the Magna ChIP HiSens Kit (Cat. # 17-10460). Successful immunoprecipitation of acetyl Histone H3 (Lys56) associated DNA fragments was verified by qPCR using ChIP Primers, human cyclin B1 promoter (Part No. CS207338).
Please refer to the MagnaChIP HiSens (Cat. # 17-10460) or EZ-MagnaChIP HiSens (Cat. # 17-10461) protocol for experimental details.

Dot Blot (Specificity) Analysis: A representative lot detected acetyl-Histone H3 (Lys56) in an Absurance Histone H3 Antibody Specificity Array (Cat. No. 16-667).

Western Blotting Analysis:
Evaluated by Western Blotting in HeLa acid extract, sodium butyrate treated HeLa acid extract, colcemid treated HeLa acid extract, recombinant Histone H3, recombinant Histone H4, and recombinant Histone H2A.
A 1:400 dilution of this antibody detected acetyl Histone H3 (Lys56) in sodium butyrate treated HeLa acid extract.
This ChIPAb+ Acetyl-Histone H3 (Lys56) -ChIP Validated Antibody & Primer Set conveniently includes the antibody & the specific control PCR primers.

Packaging

25 assays per set. Recommended use: 1 μL of antibody per chromatin immunoprecipitation (dependent upon biological context).

Quality

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from HeLa cells (1x105 cell equivalents per IP) were subjected to chromatin immunoprecipitation using 1 µL of either Normal Rabbit IgG (Part No. CS213233), or 1 µL Anti-Acetyl-Histone H3 (Lys56) (Part No.CS207337) and the Magna ChIP® HiSens Kit (Cat. # 17-10460). Successful immunoprecipitation of acetyl Histone H3 (Lys56) associated DNA fragments was verified by qPCR using ChIP Primers, human cyclin B1 promoter (Part No. CS207338).
Please refer to the MagnaChIP HiSens (Cat. # 17-10460) or EZ-MagnaChIP HiSens (Cat. # 17-10461) protocol for experimental details

Target description

~17 kDa observed

Physical form

Anti-acetyl-Histone H3 (Lys56) (Rabbit Polyclonal). One vial containing 25 µL purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH7.4),150mM NaCl, and 0.05% sodium azide, before the addition of 30% glycerol. Store at -20°C.

Normal Rabbit IgG. One vial containing 25 µL of normal rabbit IgG in buffer containing 0.01M PBS, 0.1% sodium azide, pH7.1, before the addition of 30% glycerol. Store at -20°C.

ChIP Primers, human cyclin B1 promoter. One vial containing 75 μL of 5 μM of each primer specific for human cyclin B1 promoter (chr5: 68462295-68462447, hg19 build). Store at -20°C.
FOR: 5’ CCT TGG GAA ACT GGA CAA TC 3’
REV: 5’ TTT GCC AGG GTC ACA CAT TA 3’

Analysis Note

Control
Includes normal rabbit IgG and primers specific for human cyclin B1 promoter region.

Legal Information

MAGNA CHIP is a registered trademark of Merck KGaA, Darmstadt, Germany
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wang Li et al.
Acta pharmacologica Sinica, 41(12), 1557-1567 (2020-04-09)
Sirtuin 6 (SIRT6) has been reported to play a key role in cognitive function and mood regulation, yet its role in mood disorders is not completely understood. Here, we confirmed that knockdown of hippocampal SIRT6 alleviated depression-like behaviors induced by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service