Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 mL | Check Cart for Availability | € 293,00 |
About This Item
grade
certified reference material
Quality Segment
agency
suitable for EPA 200.7, suitable for EPA 200.8
product line
Certipur®
concentration
10000 mg/L Pb in 2-3% HNO3
technique(s)
ICP: suitable
density
1.029 g/cm3 at 20 °C
application(s)
industrial qc
pharmaceutical
format
single component solution
storage temp.
15-25°C
General description
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at https://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.
Application
Other Notes
Detailed information on our complete workflow solutions for ICP-AAS can be found in our Brochure: High-Purity Chemicals for Inorganic Trace Analysis.
In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) ICP: suitable | technique(s) ICP: suitable | technique(s) ICP: suitable | technique(s) ICP: suitable |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| format single component solution | format single component solution | format single component solution | format single component solution |
| product line Certipur® | product line Certipur® | product line Certipur® | product line Certipur® |
| application(s) industrial qc | application(s) industrial qc | application(s) industrial qc | application(s) industrial qc |
| Quality Level 300 | Quality Level 300 | Quality Level 300 | Quality Level 300 |
Still not finding the right product?
Explore all of our products under Lead ICP standard
signalword
Danger
Hazard Classifications
Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Met. Corr. 1 - Repr. 1A - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
target_organs
Blood,Central nervous system,Immune system,Kidney
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
10099-74-8
10099-74-8
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.






