Skip to Content
Merck
All Photos(2)

Key Documents

169756

Sigma-Aldrich

Antithrombin III, Human Plasma

Single-chain glycoprotein present in normal serum at about 23 mg/100 ml.

Synonym(s):

Antithrombin III, Human Plasma, ATIII, Heparin cofactor, Factor Xa inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€448.00

€448.00


Estimated to ship on29 March 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€448.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

€448.00


Estimated to ship on29 March 2025


Request a Bulk Order

Quality Level

form

solid

specific activity

≥5 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

water: soluble

shipped in

ambient

storage temp.

−20°C

General description

Single chain glycoprotein present in normal serum at about 23 mg/100 ml. Principal physiological inhibitor of thrombin. Present at higher levels in plasma than in serum because it complexes with thrombin during coagulation. Functions by inhibiting proteolytic enzymes involved in blood coagulation and fibrinolysis, including Factor Xa, plasmin, thrombin, and trypsin. Potency is strongly enhanced by heparin.
Single-chain glycoprotein present in normal serum at about 23 mg/100 ml. Principal physiological inhibitor of thrombin. Present at higher levels in plasma than in serum because it complexes with thrombin during coagulation. Functions by inhibiting proteolytic enzymes involved in blood coagulation and fibrinolysis, including Factor Xa, plasmin, thrombin, and trypsin. Potency is strongly enhanced by heparin.

Biochem/physiol Actions

Cell permeable: no
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of antithrombin that will inhibit 1.0 unit of human plasma thrombin present in excess. Thrombin activity is measured using 500 µM tosyl-Gly-Pro-Arg-pNA as substrate in 100 mM Tris-HCl at 25°C, pH 8.2

Physical form

Lyophilized from 100 mM sodium citrate buffer, 20 mM Tris-HCl, pH 8.3.

Preparation Note

Please refer to label for lot-specific volume prior to lyophilization.

Analysis Note

single band by SDS-PAGE

Other Notes

Tsiang, M., et al. 1997. J. Biol. Chem. 272, 12024.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service