Skip to Content
Merck
All Photos(1)

Key Documents

1.67730

Supelco

Continous level sensor for organic solvents in 30l stainless steel barrels

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€6,260.00

€6,260.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
1 UNIT
€6,260.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.03

€6,260.00


Estimated to ship on19 May 2025


Request a Bulk Order

storage temp.

no temp limit

Related Categories

General description

Continuous level sensor technology with ATEX approval provides maximum safety and reliability for your daily work. The graphical display always shows the exact solvent level inside the container. With adjustable alarm, individual management, measurement and control of solvent supply and disposal now are available and automatable, e.g. by visible control alarms or automatical control of HPCL instruments.
incl. G2 threaded adapter, dip tube, graphical display and cables (length: 5m)

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service