Skip to Content
Merck

Skip To

1.66702

Sabouraud Dextrose Agar

with Lecithin, Tween®, Histidine and Sodium thiosulfate - 90mm ICR+ Settle plate, irradiated, triple packed, for environmental monitoring (Isolator and Clean room)

Synonym(s):

SDA, ICR plus Settle plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41106227
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polystyrene plate

Quality Segment

agency

EP, USP

sterility

sterile; γ-irradiated

form

plate

feature

2D bar code, ready-to-use

packaging

pkg of (20 Plates per box)

D

90 mm

pH

(7.3 (in H2O))

application(s)

air monitoring
microbiology
pharmaceutical

storage temp.

15-25°C

suitability

(Yeasts and Molds)

General description

Sabouraud Dextrose Agar + LTHTh - ICR plus settle plate is a selective media for the determination of the total count of yeasts and molds. Settle plates are used for active or passive air monitoring as well for fingerprints of personnel. Sabouraud Dextrose Medium contains digests of animal tissues (peptones) which provide a nutritious source of amino acids and nitrogenous compounds for the growth of fungi and yeasts. Dextrose is added as the energy and carbon source and low pH favours fungal growth. In addition, Lecithin, polysorbate 80 (Tween®) Histidine and Sodium thiosulfate are neutralizers to inactivate residues of disinfectants. In addition, the plate model (plus or “+”) is supplied with a lockable lid for safe transport of the samples without the risk of losing the lid. For anaerobic incubation the lid needs to be turned into the vent position.

Application

Sabouraud Dextrose Agar (SDA) is a selective medium for cultivation and isolation of yeasts and molds. The media is used for environmental monitoring by determining the yeast and mold count in air by active or passive air monitoring as well as fingerprints of personnel.

Packaging

1667020020-CN: 20 Plates per box

Legal Information

TWEEN is a registered trademark of Croda International PLC


Still not finding the right product?

Explore all of our products under Sabouraud Dextrose Agar


Storage Class

13 - Non Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters