Skip to Content
Merck
All Photos(1)

Key Documents

165350

Sigma-Aldrich

PARP Inhibitor I, 3-ABA

The PARP Inhibitor I, 3-ABA, also referenced under CAS 3544-24-9, controls the biological activity of PARP. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

PARP Inhibitor I, 3-ABA, 3-Aminobenzamide

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€60.80

€60.80


Estimated to ship on28 May 2025



Select a Size

Change View
100 MG
€60.80

About This Item

Empirical Formula (Hill Notation):
C7H8N2O
CAS Number:
Molecular Weight:
136.15
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€60.80


Estimated to ship on28 May 2025


description

RTECS - CU8992000

Quality Level

Assay

≥99% (GC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

ethanol: 1 mg/mL
water: 1 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

Nc1cc(ccc1)C(=O)N

InChI

1S/C7H8N2O/c8-6-3-1-2-5(4-6)7(9)10/h1-4H,8H2,(H2,9,10)

InChI key

GSCPDZHWVNUUFI-UHFFFAOYSA-N

General description

A cell-permeable anti-proliferative agent that acts as an inhibitor of poly(ADP-ribose) polymerase (PARP). Has minimal effect on bacterial toxin-mediated ADP-ribosylation. An inhibitor of UV-induced apoptosis. Also strongly inhibits K562 target cell killing by human effector natural killer cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PARP
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C ). Stock solutions are stable for 3 months at -20°C.

Other Notes

Tiozzo, R., et al. 1996. Biochem. Biophys. Res. Commun. 225, 826.
Malorni, W., et al. 1995. Biochem. Biophys. Res. Commun. 207, 715.
Malorni, W., et al. 1994. Biochem. Biophys. Res. Commun. 199, 1250.
Monti, D., et al. 1994. Biochem. Biophys. Res. Commun.199, 525.
Kozak, K.R., and Ross, I.A. 1991. Biochem. Biophys. Res. Commun.179, 1225.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service