Skip to Content
Merck
All Photos(1)

Key Documents

1.52370

Supelco

Chromolith® WP 300 RP-18 (monolithic) HPLC Columns

L × I.D. 10 cm × 2 mm HPLC Column

Synonym(s):

Chromolith® WP 300 RP-18 100-2 HPLC column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€1,220.00

€1,220.00


Estimated to ship on19 May 2025



Select a Size

Change View
1 EA
€1,220.00

About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

€1,220.00


Estimated to ship on19 May 2025


Product Name

Chromolith® WP 300 RP-18, L × I.D. 10 cm × 2 mm HPLC column

material

PEEK column

Quality Level

Agency

suitable for USP L1

product line

Chromolith®

feature

endcapped: no

packaging

1 ea of

extent of labeling

18% loading

parameter

200 bar pressure
60 °C max. temp.

technique(s)

HPLC: suitable
LC/MS: suitable
UHPLC-MS: suitable
UHPLC: suitable
mass spectrometry (MS): suitable

column L × I.D.

10 cm × 2 mm

surface area

300 m2/g

matrix

monolithic silica

matrix active group

C18 bonding phase

pore size

2 μm macroporosity
30 mm mesoporosity

operating pH range

2-7.5

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

The most hydrophobic of the Chromolith® WP 300 line, the RP-18 column is useful for the resolution of peptides and smaller proteins. One critical quality attribute (CQA) required by regulatory bodies is the peptide map of a biotherapeutic. Peptide maps generated by RP-HPLC provide valuable information about protein structure, stability, and purity. To be effective, the RPHPLC column must be able to resolve a high percentage of the peptides in the sample. The more peptides, the better the information.
The Chromolith® WP 300 RP-18 column gives unsurpassed RP-HPLC resolution of peptide maps from enzymatic digests. The improvements in silica and bonded-phase chemistry incorporated into the Chromolith® WP 300 line improve resolution by increasing efficiency and reducing peak tailing.

Chromolith® WP 300 columns are made of monolithic silica with a characteristic bimodal pore structure.

Chromolith® WP 300 RP-18, 2 mm I.D. columns have macropores with 2 μm diameter, giving a column efficiency exceeding 80,000 plates/meter. The mesopores are 30 nm (300 Å), the surface modification is Octadecylsilane. Chromolith® WP 300 RP-18 columns fall under the USP L1 Classification. Chromolith® WP 300 RP-18 columns are fully suitable for LC-MS applications.
The column dimension of 100 mm length and 2 mm I.D. is the best choice for fast separations of Biomolecules on any UHPLC System.

Chromolith® WP 300 RP-18, 100-2 HPLC columns enable:
—Completely bioinert column hardware and high biorecovery
—High-speed separations at very low column back-pressure
—High matrix tolerance, allowing time and cost savings with simplified workflows
—Cost savings due to extended column lifetimes
—Connection of columns, flow gradients and the use of mobile phases with higher viscosity

The recommended Guard-columns are:
Chromolith® WP 300 RP-18, 5-2 guard cartridges (3 pieces) [1.52372] with the Chromolith® Guard Cartridge Holder Bioinert 5 mm x 2 mm [1.52355]

The Chromolith® column coupler can be used to significantly increase the separation efficiency of Chromolith® columns by coupling several columns in series. [1.51467]

Learn more:
Brochure: Race through your Separations on Any System. Chromolith® Monolithic Silica HPLC Columns

All Supelco HPLC Columns, including monolithic Chromolith® columns, are fully compatible to all HPLC and UHPLC Instruments.
Supelco HPLC Products: The Best Choice for Any LC Instrument

Legal Information

Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Compare columns in resolving medium-sized antibody fragments after digestion with DTT or IdeS using Reversed-Phase Chromatography for analysis.

Intact analysis of Universal Antibody Standard human and Antibody-Drug Conjugate (ADC) Mimic on silica monolithic HPLC columns under various conditions.

Related Content

Analytical Chromolith® HPLC columns for Bio applications, 300A mesopores: RP-18, RP-8, RP-4, Prot-A, Epoxy

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service