Skip to Content
Merck
All Photos(1)

Key Documents

1.52350

Supelco

Chromolith® WP 300 Epoxy (monolithic) HPLC Columns

L × I.D. 10 cm × 2 mm HPLC Column

Synonym(s):

Chromolith® WP 300 Epoxy 100-2 HPLC column

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€1,080.00

€1,080.00


Estimated to ship on17 May 2025



Select a Size

Change View
1 PKG
€1,080.00

About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

€1,080.00


Estimated to ship on17 May 2025


Product Name

Chromolith® WP 300 Epoxy, L × I.D. 10 cm × 2 mm HPLC column

material

PEEK column

Quality Level

product line

Chromolith®

feature

endcapped: no

packaging

1 ea of

extent of labeling

5-6% Carbon loading

parameter

2-60 °C temp. range
200 bar pressure

technique(s)

HPLC: suitable
LC/MS: suitable
UHPLC-MS: suitable
UHPLC: suitable

column L × I.D.

10 cm × 2 mm

matrix

monolithic silica

matrix active group

Epoxy bonding phase

pore size

2 μm macroporosity
30 nm mesoporosity

operating pH range

1.5-7.5

Looking for similar products? Visit Product Comparison Guide

General description

Chromolith® WP 300 Epoxy columns are specially designed for the user-specific immobilization of ligands and their later application in HPLC. The unique bimodal pore structure of silica monoliths allows efficient coupling independent of molecule size. The wider mesopores also enable the use of proteins and antibodies as both ligand immobilized on the column, and later analyte separated by an immobilized column.
Potential applications: attach trypsin to obtain HPLC column-protein digestion reactor; attach protein and measure other protein interaction with the attached one; attach any chiral selector to obtain a chiral column, attach any affinity ligand to obtain custom made affinity column, etc.

Chromolith® WP 300 Epoxy 100-2 columns are made of monolithic silica with a characteristic bimodal pore structure.

Key Benefits:
—Easy to perform immobilization of ligand opens up limitless customization options
—Monolithic format allows for high flow rates at low backpressure, enabling high throughput
—Narrow column I.D. enables improved performance and improved MS-compatibility

Learn more:
Brochure: Race through your Separations on Any System. Chromolith® Monolithic Silica HPLC Columns

Flyer: [Create Your Own Column Chemistry - Chromolith® WP 300 Epoxy 2 mm I.D. HPLC Columns]

All Supelco HPLC Columns, including monolithic Chromolith® columns, are fully compatible to all HPLC and UHPLC Instruments.
Supelco HPLC Products: The Best Choice for Any LC Instrument

Legal Information

Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Analytical Chromolith® HPLC columns for Bio applications, 300A mesopores: RP-18, RP-8, RP-4, Prot-A, Epoxy

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service