Skip to Content
Merck
All Photos(1)

Key Documents

14-939-M

Sigma-Aldrich

ErbB2, active

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€580.00

€580.00


Estimated to ship on24 May 2025



Select a Size

Change View
10 μG
€580.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

€580.00


Estimated to ship on24 May 2025


biological source

human

Quality Level

mol wt

Mw 69 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

General description

Protein tyrosine kinase that is part of several cell surface receptor complexes, but that apparently needs a coreceptor for ligand binding. Essential component of a neuregulin-receptor complex, although neuregulins do not interact with it alone. GP30 is a potential ligand for this receptor. Regulates outgrowth and stabilization of peripheral microtubules (MTs). Upon ERBB2 activation, the MEMO1-RHOA-DIAPH1 signaling pathway elicits the phosphorylation and thus the inhibition of GSK3B at cell membrane. This prevents the phosphorylation of APC and CLASP2, allowing its association with the cell membrane. In turn, membrane-bound APC allows the localization of MACF1 to the cell membrane, which is required for microtubule capture and stabilization

Application

Research Category
Signaling

Biochem/physiol Actions

Protein Target: ErbB2
Target Sub-Family: TK

Packaging

Also available in 250μg size as catalog # 14-939M and in bulk as catalog # 14-939-K.

Quality

Routinely evaluated by phosphorylation of poly(Glu, Tyr) (4:1)

Physical form

Protein in 50mM Tris/HCl pH7.5, 150mM NaCl, 0.5mM EDTA, 0.01% CHAPS, 50% glycerol, 2mM DTT. Frozen solution.
Purified using immobilized metal affinity chromatography.

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service