Skip to Content
Merck
All Photos(1)

Key Documents

14-937-M

Sigma-Aldrich

ALK2, active

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.28

recombinant

expressed in baculovirus infected Sf21 cells

Quality Level

purified by

immobilized metal affinity chromatography (IMAC)

technique(s)

activity assay: suitable

UniProt accession no.

Gene Information

human ... ACVR1(90)

General description

Activins are dimeric growth and differentiation factors which belong to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. Activins signal through a heteromeric complex of receptor serine kinases which include at least two type I ( I and IB) and two type II (II and IIB) receptors. These receptors are all transmembrane proteins, composed of a ligand-binding extracellular domain with cysteine-rich region, a transmembrane domain, and a cytoplasmic domain with predicted serine/threonine specificity. Type I receptors are essential for signaling; and type II receptors are required for binding ligands and for expression of type I receptors. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. This gene encodes activin A type I receptor which signals a particular transcriptional response in concert with activin type II receptors. Mutations in this gene are associated with fibrodysplasia ossificans progressive.

Packaging

Also available in 10μg size as catalog # 14-937 and in bulk as catalog # 14-937-K.
Also available in 250μg size as catalog # 14-937M and in bulk as catalog # 14-937-K.

Quality

Routinely evaluated by phosphorylation of casein

Physical properties

45 kDa

Physical form

Protein in 50mM Tris/HCl pH7.5, 300mM NaCl, 0.1mM EGTA, 0.03% Brij-35, 270mM sucrose, 1mM benzamidine, 0.2mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.

Storage and Stability

6 months at -70°C

Analysis Note

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service