Skip to Content
Merck
All Photos(1)

Key Documents

14-835-M

Sigma-Aldrich

Aurora B Protein, active, 10 µg

Active, His-tagged, recombinant, human Aurora B, full length & N-terminal GST-tagged, recombinant, human INCENP, amino acids 821-end, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€581.00

€581.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
€581.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

€581.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 38 kDa (INCENP)
Mw 40 kDa (Aurora B)

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

General description

His-tagged, recombinant, human Aurora B, full length and N-terminal GST-tagged, recombinant, human INCENP, amino acids 821-end
Product Source: human Aurora B, human INCENP co-expressed by baculovirus in Sf21 insect cells.

Application

Research Category
Apoptosis & Cancer

Biochem/physiol Actions

Protein Target: Aurora-B
Target Sub-Family: AGC

Physical form

Glutathione-agarose

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service