Skip to Content
Merck
All Photos(1)

Key Documents

14-781-M

Sigma-Aldrich

PI3 Kinase (p110α(E545K)/p85α), Murine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf21 cells

Quality Level

100
300

description

protein target PI3Kα/p85α

purified by

(Ni2+/NTA-agarose)

technique(s)

activity assay: suitable

UniProt accession no.

Gene Information

mouse ... PIK3CA(18706)

General description

FUNCTION: SwissProt: P42336 # Phosphorylates PtdIns, PtdIns4P and PtdIns(4,5)P2 with a preference for PtdIns(4,5)P2.
SIZE: 1068 amino acids; 124284 Da
SUBUNIT: Heterodimer of a p110 (catalytic) and a p85 (regulatory) subunit. Binds to IRS1 in nuclear extracts. Interacts with RUFY3 (By similarity).
DISEASE: SwissProt: P42336 # Defects in PIK3CA are associated with colorectal cancer (CRC) [MIM:114500]. & Defects in PIK3CA are associated with breast cancer [MIM:114480]. & Defects in PIK3CA are associated with epithelial ovarian cancer (EOC) [MIM:604370].
SIMILARITY: SwissProt: P42336 ## Belongs to the PI3/PI4-kinase family. & Contains 1 C2 domain. & Contains 1 PI3K/PI4K domain.

Packaging

Also available in 250 μg size --call for pricing and availability and reference catalog number 14-781M when ordering the 250 μg size.
Also available in 10 μg size (cat#14-781) and in bulk (cat# 14-781-K).

Physical properties

p110α (Ε545Κ) MW = 129kDa,
p85α MW = 83.6kDa.

Storage and Stability

Store at -70°C from date of shipment. For maximum recovery of product, centrifuge original vial prior to removing the cap. Rapidly thaw the vial under cold water and immediately place on ice. Aliquot unused material into pre-chilled microcentrifuge tubes and immediately snap-freeze the vials in liquid nitrogen prior to re-storage at -70°C.

Analysis Note

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service