biological source
human
Quality Level
100
300
mol wt
Mw 85.2 kDa
manufacturer/tradename
Upstate®
technique(s)
activity assay: suitable (kinase)
NCBI accession no.
UniProt accession no.
General description
6His-tagged, recombinant, full length, human Rsk4
expressed by baculovirus in Sf21 cells
Application
Active, 6His-tagged, recombinant, full length, human Rsk4, for use in Kinase Assays.
Biochem/physiol Actions
- Protein Target: Rsk4
- Target Sub-Family:AGC
Quality
routinely evaluated by phosphorylation of PAKtide (RRRLSFAEPG)
Physical form
Format: Ni2+/NTA-agarose
Storage and Stability
6 months at -70°C from date of shipment
Other Notes
For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service