Skip to Content
Merck
All Photos(1)

Key Documents

14-687-M

Sigma-Aldrich

p38α/SAPK2a (T106M) Protein, active, 10 µg

Active, N-terminal GST-tagged, recombinant, human, full-length p38α/SAPK2a containing the mutation T106M. Activated using DD-MKK6 & repurified using glutathione-agarose. For use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€545.00

€545.00


Estimated to ship on30 April 2025



Select a Size

Change View
10 μG
€545.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

€545.00


Estimated to ship on30 April 2025


biological source

human

Quality Level

100
300

recombinant

expressed in E. coli

mol wt

Mw 67.7 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

UniProt accession no.

Gene Information

human ... MAPK14(1432)

General description

N-terminal GST-tagged, recombinant, human, full-length p38α/SAPK2a containing the mutation T106M. Activated using DD-MKK6 (14-537) and repurified using glutathione- agarose.

Contains the gatekeeper mutation T160M in the ATP-binding domain. This mutation increases resistance to SB203580 as compared to the wild type sequence.
Product Source: Human, full-length p38α/SAPK2a containing the mutation T106M. Expressed in E. coli.

Quality

routinely evaluated by phosphorylation of MBP (13-104).

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service