Skip to Content
Merck
All Photos(1)

Key Documents

14-683-M

Sigma-Aldrich

PAK3 Protein, active, 10 µg

Active, C-terminal 6His-tagged, recombinant, human full length, PAK3. For use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€436.00

€436.00


Estimated to ship on23 April 2025



Select a Size

Change View
10 μG
€436.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

€436.00


Estimated to ship on23 April 2025


biological source

human

Quality Level

100
300

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 63.2 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... PAK3(5063)

General description

C-terminal 6His-tagged, recombinant, human full length, PAK3.

mitosis
Product Source: PAK3 expressed by baculovirus in Sf21 cells.

Application

Research Category
Inflammation & Immunology

Biochem/physiol Actions

Protein Target: PAK3
Target Sub-Family: STE

Quality

routinely evaluated by phosphorylation of PAKtide (RRRLSFAEPG).

Physical form

Ni2+/NTA-agarose

Storage and Stability

6 months at -70°C from date of shipment

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Roig et al.
Vitamins and hormones, 62, 167-198 (2001-05-11)
The p21-activated protein kinase gamma-PAK, also known as PAK2, has very different properties from the other two highly conserved isoforms of the PAK family, alpha-PAK (PAK1) and beta-PAK (PAK3). gamma-PAK has cytostatic activity, as shown by inhibition of cleavage of
Rho proteins, mental retardation and the neurobiological basis of intelligence.
van Galen, Elly J M and Ramakers, Ger J A
Progress in Brain Research, 147, 295-317 (2005)
Ger J A Ramakers
Trends in neurosciences, 25(4), 191-199 (2002-05-10)
For several decades, it has been known that mental retardation (MR) is associated with abnormalities in dendrites and dendritic spines. The recent cloning of seven genes that cause nonspecific MR when mutated provides important insights in the cellular mechanisms that
Zahara M Jaffer et al.
The international journal of biochemistry & cell biology, 34(7), 713-717 (2002-04-16)
The p21-activated kinases (Paks) are serine/threonine protein kinases that bind to and, in some cases, are stimulated by activated forms of the small GTPases, Cdc42 and Rac. With the recent discovery of several novel isoforms, Paks are now categorized into

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service