Skip to Content
Merck
All Photos(1)

Documents

1.46785

Millipore

Tryptic Soy Agar - Ready-to-use Settle Plates

IsoBag®, Tween®, Lecithin, DPTE Beta Bag with 190 mm beta-port, ICR plus lockable plate, sterile; irradiated, plate diam. 90 mm, single packed of, suitable for air monitoring

Synonym(s):

IsoBag® Tryptic Soy Agar Settle Plus, TSA, Soybean Casein Digest Agar, Isolator Settle plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; irradiated

form

plate

feature

rapid transfer port

composition

Lecithin
Tween®

manufacturer/tradename

IsoBag®

packaging

single packed of

plate diam.

90 mm

application(s)

air monitoring
pharmaceutical

compatibility

for use with MAS-100®

suitability

bacteria
molds
yeasts

Looking for similar products? Visit Product Comparison Guide

General description

The IsoBag® Tryptic Soy Agar settle plate is designed for quick and convenient transfer of culture media for air monitoring in isolators or RAB systems. The IsoBag® consists of a polyurethane DPTE Beta Bag compatible with 190 mm Alpha Ports. The bag contains 10 stacks (single wrapped) of settle plates and sterile Polyethylene zip lock bags to mitigate the risk of a cross contamination during transport of the plates. Tryptic Soy Agar is supplemented with Lecithin, polysorbate (Tween®) 80 and pyruvate to neutralize residues of disinfectants. In addition, the plate model (plus or “+”) is supplied with a lockable lid for safe transportation. The IsoBag® is designed to reduce the decontamination cycles in production isolators.

Application

The IsoBag® TSA Settle plate plus is a sterile polyurethane DPTE Beta Bag designed for quick and convenient transfer of culture media for air monitoring in isolators or RAB systems

Legal Information

ISOBAG is a registered trademark of Merck KGaA, Darmstadt, Germany
MAS-100 is a registered trademark of MBV AG
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

IsoBag® ease the environmental monitoring workflow in aseptic production isolator

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service