Skip to Content
Merck
All Photos(1)

Key Documents

1.46720

Millipore

Tryptic Soy Agar - Ready-to-use Contact Plates

Thiosulphate, Lecithin, Histidine, Tween®, ICR Plate with non-lockable lid, plate diam. 55 mm, sterile; irradiated, suitable for surface monitoring

Synonym(s):

CASO Agar, Casein Soybean Digest Agar with Lecithin, Tween, Histidine andThiosulfate, TSA, Tryptic Soy Agar, Tryptic Soy Agar with neutralizer, Tryptone Soya Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.74
Pricing and availability is not currently available.

Quality Level

sterility

sterile; irradiated

form

solid

feature

ready-to-use

composition

Casein Soybean Digest Agar with Lecithin, Tween, Histidine and Thiosulfate
Histidine
Lecithin
Thiosulphate
Tween®

packaging

single packed of

technique(s)

Rapid Sterility Testing: suitable

plate diam.

55 mm

application(s)

pharmaceutical
surface monitoring

compatibility

stand alone

suitability

aerobic bacteria
anaerobic bacteria

Looking for similar products? Visit Product Comparison Guide

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service