Skip to Content
Merck
All Photos(1)

Key Documents

1.46461

Millipore

MacConkey Broth

ready-to-use, bottle filling volume 100 mL

Synonym(s):

Broth Medium G, MacC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74
Pricing and availability is not currently available.

Agency

EP 2.6.1
JP 4.06
USP 71

Quality Level

sterility

sterile; autoclaved

form

liquid

shelf life

12 mo.

feature

closure type blue screw cap with 3 loci
ready-to-use

packaging

single packed of 10 bottles
bottle of

technique(s)

direct inoculation: suitable
sterile filtration: suitable

bottle capacity

(125 mL)

bottle filling volume

100 mL

capacity

125 mL

pH

7.3

application(s)

bioburden testing
cosmetics
food and beverages
pharmaceutical

storage temp.

2-25°C

suitability

Escherichia coli
enterobacteriaceae

Looking for similar products? Visit Product Comparison Guide

General description

The MacConkey Broth supports the growth of most gram-negative bacteria, whereas the presence of bile salts inhibits the growth of gram-positive bacteria. The color of the medium is changed from violet to yellow due to the degradation of lactose to acid by bacteria. The change in pH is indicated by the pH indicator bromocresol purple, which shows reduced inhibition compared to neutral red.

Application

MacConkey Broth is suitable for the sterility testing of pharmaceutical products. It is designed for the enrichment of Enterobacteriaceae and the detection of Escherichia coli.

Features and Benefits

  • Ready-to-use media
  • Bottles with screw caps

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service