Skip to Content
Merck

Skip To

14-616-M

MSK2 Protein, active, 10 µg

Active, N-terminal His6-tagged, recombinant, human MSK2, amino acids 2-end, for use in Kinase Assays.

Promotion Available

Sign In to View Organizational & Contract Pricing.

Spend € 450 — Save 5 %; Spend € 700 — Save 10 %; Spend € 1.500 — Save 15 %*

Sign in to check eligibility. Shop Products

Select a Size

Change View
Size/SKUAvailabilityPrice
10 μg
Check Cart for Availability
€ 483,00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
eCl@ss:
32160405
Biological source:
human
Recombinant:
expressed in baculovirus infected Sf21 cells
Mol wt:
Mw 89.9 kDa

€ 483,00

Save Up To 15 %


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 89.9 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... RPS6KA4(8986)

General description

N-terminal His6-tagged, recombinant, human MSK2, amino acids 2-end

Biochem/physiol Actions

Protein Target: MSK2

Physical form

Purified using Ni2+/NTA-agarose. Activated using MAP Kinase 2/Erk2, active (Catalog # 14-173) and repurified using Ni2+/NTA-agarose

Analysis Note

routinely evaluated by phosphorylation of 30mM Crosstide (Catalog # 12-331)

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters