Skip to Content
Merck
All Photos(1)

Key Documents

14-459-M

Sigma-Aldrich

Abl Protein, active, mouse, 10 µg

Active, recombinant mouse Abl, residues 27-1123, containing N-terminal His-6 tag, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€462.00

€462.00


Estimated to ship on16 May 2025



Select a Size

Change View
10 μG
€462.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

€462.00


Estimated to ship on16 May 2025


biological source

mouse

Quality Level

100
300

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 123.5 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

General description

Product Source: Expressed by baculovirus in Sf21
Recombinant mouse Abl, residues 27-1123, containing N-terminal His6 tag

Biochem/physiol Actions

Protein Target: Abl
Target Sub-Family: TK

Packaging

Also available in 250 µg size --call for pricing and availability and reference catalog number 14-459M when ordering the 250 µg size.

Quality

routinely evaluated by phosphorylation of Abltide

Physical form

Ni2+/NTA-agarose

Storage and Stability

1 year at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Activation of the c-Abl tyrosine kinase in the stress response to DNA-damaging agents.
Kharbanda, S, et al.
Nature, 376, 785-788 (1995)
B B Brasher et al.
The Journal of biological chemistry, 275(45), 35631-35637 (2000-08-31)
Using the specific Abl tyrosine kinase inhibitor STI 571, we purified unphosphorylated murine type IV c-Abl and measured the kinetic parameters of c-Abl tyrosine kinase activity in a solution with a peptide-based assay. Unphosphorylated c-Abl exhibited substantial peptide kinase activity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service