14-343
FKHR Protein, 100 µg
N-terminal, Glutathione-Stransferase (GST) tagged, full-length fusion protein corresponding to human FKHR.
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
biological source
human
mol wt
Mw 105 kDa
manufacturer/tradename
Upstate®
technique(s)
activity assay: suitable (kinase)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
Gene Information
human ... FOXO1(2308)
General description
Product Source: Recombinant human expressed in E. coli
Application
N-terminal, Glutathione-Stransferase (GST) tagged, full-length fusion protein corresponding to human FKHR.
Quality
routinely evaluated by phosphorylation by Akt1
Physical form
Glutathione-agarose
Storage and Stability
1 year at -70°C
Other Notes
For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Scientific reports, 5, 14527-14527 (2015-09-30)
The formation and maintenance of connectivity are critically important for the processing and storage of information in neuronal networks. The brain extracellular matrix (ECM) appears during postnatal development and surrounds most neurons in the adult mammalian brain. Importantly, the removal
Journal of neurophysiology, 107(2), 640-648 (2011-10-28)
The midbrain is an important processing area for sensory information in vertebrates. The optic tectum and its mammalian counterpart, the superior colliculus, receive multimodal, topographic information and contain a sensory map that plays a role in spatial attention and orientation
Neuron, 104(6), 1153-1167 (2019-11-02)
Anatomical and behavioral data suggest that the ventrolateral orbitofrontal cortex (VLO), which exhibits extensive connectivity and supports diverse sensory and cognitive processes, may exert global influence over brain activity. However, this hypothesis has never been tested directly. We applied optogenetic
Molecular and cellular biology, 26(1), 50-62 (2005-12-16)
The GTPase-deficient, activated mutant of Galpha12 (Galpha12Q229L, or Galpha12QL) induces neoplastic growth and oncogenic transformation of NIH 3T3 cells. Using microarray analysis, we have previously identified a role for platelet-derived growth factor receptor alpha (PDGFRalpha) in Galpha12-mediated cell growth (R.
Journal of neurophysiology, 123(2), 815-830 (2020-01-23)
The interplay between inhibition and excitation can regulate behavioral expression and control, including the expression of communicative behaviors like birdsong. Computational models postulate varying degrees to which inhibition within vocal motor circuitry influences birdsong, but few studies have tested these
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service