Skip to Content
Merck
All Photos(1)

Key Documents

14-327-M

Sigma-Aldrich

JNK1α1/SAPK1c Protein, active, 10 µg

Active, recombinant full-length human JNK1α1/SAPK1c with an N-terminal His-tag, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€547.00

€547.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
€547.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

€547.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in Sf21 cells

product line

Upstate®

specific activity

(For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.)

mol wt

Mw 45 kDa

purified by

(Ni2+/NTA-agarose)

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

accession no.

 (FUNCTION: SwissProt: P45983 # JNK1 isoforms display different binding patterns: beta-1 preferentially binds to c-Jun, whereas alpha-1, alpha-2, and beta- 2 have a similar low level of binding to both c-Jun or ATF2. However, there is no correlation between binding and phosphorylation, which is achieved at about the same efficiency by all isoforms. )
NM_002750.2

UniProt accession no.

Gene Information

human ... MAPK8(5599)

General description

Recombinant full-length human JNK1α1/SAPK1c with an N-terminal His-tag

Packaging

10 μg vial
Also available in 250μg size (catalog number 14-327M).
Also available in 10 μg size and in bulk

Quality

Routinely evaluated by phosphorylation of ATF2 (amino acids 19-96)

Target description

JNK1α1/SAPK1c

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Girish V Putcha et al.
Neuron, 38(6), 899-914 (2003-06-24)
Trophic factor deprivation (TFD) activates c-Jun N-terminal kinases (JNKs), culminating in coordinate AP1-dependent transactivation of the BH3-only BCL-2 proteins BIM(EL) and HRK, which in turn are critical for BAX-dependent cytochrome c release, caspase activation, and apoptosis. Here, we report that
Y Zhang et al.
The Journal of biological chemistry, 276(24), 20913-20923 (2001-03-30)
Ultraviolet light A (UVA) plays an important role in the etiology of human skin cancer, and UVA-induced signal transduction has a critical role in UVA-induced skin carcinogenesis. The upstream signaling pathways leading to p70(S6K) phosphorylation and activation are not well
S Gupta et al.
Science (New York, N.Y.), 267(5196), 389-393 (1995-01-20)
Treatment of cells with pro-inflammatory cytokines or ultraviolet radiation causes activation of the c-Jun NH2-terminal protein kinase (JNK). Activating transcription factor-2 (ATF2) was found to be a target of the JNK signal transduction pathway. ATF2 was phosphorylated by JNK on
S Gupta et al.
The EMBO journal, 15(11), 2760-2770 (1996-06-03)
The JNK protein kinase is a member of the MAP kinase group that is activated in response to dual phosphorylation on threonine and tyrosine. Ten JNK isoforms were identified in human brain by molecular cloning. These protein kinases correspond to
Y Fleming et al.
The Biochemical journal, 352 Pt 1, 145-154 (2000-11-04)
Stress-activated protein kinase 1 (SAPK1), also called c-Jun N-terminal kinase (JNK), becomes activated in vivo in response to pro-inflammatory cytokines or cellular stresses. Its full activation requires the phosphorylation of a threonine and a tyrosine residue in a Thr-Pro-Tyr motif

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service