Skip to Content
Merck
All Photos(1)

Key Documents

14-251-M

Sigma-Aldrich

p38α/SAPK2a Protein, active, 10 µg

Active, N-terminal GST tagged, recombinant, full length p38α/SAPK2a, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€547.00
250 μG
€3,470.00

€547.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
€547.00
250 μG
€3,470.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

€547.00


Estimated to ship on18 May 2025


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in E. coli

mol wt

Mw 67.7 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

UniProt accession no.

Gene Information

human ... MAPK14(1432)

General description

N-terminal GST tagged, recombinant, full length p38α/SAPK2a
Product Source: expressed in E. coli

Biochem/physiol Actions

Protein Target: p38α/SAPK2a
Target Sub-Family: CMGC

Packaging

Also available in 250μg size (2x125μg) - Call for pricing and availability. Please reference catalog number 14-251M when ordering the 250μg size.

Quality

Routinely evaluated by phosphorylation of MBP

Physical form

Glutathione-agarose

Storage and Stability

6 months at -20°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y N Doza et al.
FEBS letters, 364(2), 223-228 (1995-05-08)
A MAP kinase homologue, termed the reactivating kinase (RK), lies in a signalling pathway which mediates cellular responses to stress. Here we demonstrate that the stress-induced activation of the RK in human KB cells is accompanied by the phosphorylation of
A single autophosphorylation site confers oncogenicity to the Neu/ErbB-2 receptor and enables coupling to the MAP kinase pathway.
Ben-Levy, R, et al.
The Embo Journal, 13, 3302-3311 (1994)
A Cuenda et al.
FEBS letters, 364(2), 229-233 (1995-05-08)
A class of pyridinyl imidazoles inhibit the MAP kinase homologue, termed here reactivating kinase (RK) [Lee et al. (1994) Nature 372, 739-746]. We now show that one of these compounds (SB 203580) inhibits RK in vitro (IC50 = 0.6 microM)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service