Skip to Content
Merck
All Photos(1)

Documents

1.40143

Millipore

Tryptic Soy Broth - Dehydrated Culture Media

EcoCult®, EP, USP, JP, ISO 11133, FDA (BAM), USDA-FSIS, APHA, powder, suitable for bioburden testing, microbiology

Synonym(s):

TSB, CASO, Casein Soybean Digest Broth, Casein-Peptone Soymeal-Peptone Broth, Tryptone Soya Broth, SCDM, CASO Broth, Casein Soya Broth, Soybean Casein digest Broth, TSB, Tryptone Soya Broth

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106213
NACRES:
NA.74

Agency

APHA
EP
ISO 11133
JP
USDA-FSIS
USP

Quality Level

reg. compliance

FDA (BAM)

sterility

non-sterile

form

powder

manufacturer/tradename

EcoCult®

technique(s)

microbiological culture: suitable

application(s)

bioburden testing
microbiology

food and beverages
pathogen testing
pharmaceutical

storage temp.

10-30°C

suitability

Cronobacter spp. (spp.)
Escherichia coli (pathogenic)
Salmonella spp. (spp.)
anaerobic bacteria

Looking for similar products? Visit Product Comparison Guide

General description

EcoCult® Tryptic Soy Broth is a universal culture medium free from inhibitors and indicators. The medium is suitable for the isolation and cultivation of fastidious aerobic bacteria, yeasts, and molds. The combination of the two peptones, enzymatic digest of casein and of soybean, provides a high nutrition by supplying organic nitrogen, amino acids, and longer-chained peptides. Sodium chloride supplies the osmotic balance to this complex medium, while the dipotassium phosphate helps in buffering.

Application

EcoCult® Tryptic Soy Broth is suitable for the isolation and cultivation of a wide range of microorganisms from different material. It is used for sterility testing and microbial enumeration of the non-sterile preparations and products. It is used for the determination of total viable aerobic count using the MPN method. It is also used as pre-enrichment medium for the detection of food pathogens, e.g. Salmonella, STEC.

Features and Benefits

  • Compliance to major regulatory standards ISO, FDA-BAM, EPA or USP
  • Clear labeling and Certificate of Analysis to check compliance at a glance
  • Reliable and accurate quality in powder format
  • Cost-effective pricing

Legal Information

ECOCULT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service