Skip to Content
Merck
All Photos(2)

Key Documents

1.40006

Supelco

Extran® AP 17

greener alternative

liquid, alkaline concentrate

Synonym(s):

TM=["EXTRAN"] AP 17 cleaning agent; TM=["EXTRAN"] AP 17 detergent

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 L
€76.30
5 L
€130.00
5 L
€130.00
10 L
€229.00
25 L
€458.00

€76.30


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
2.5 L
€76.30
5 L
€130.00
5 L
€130.00
10 L
€229.00
25 L
€458.00

About This Item

UNSPSC Code:
41103206
NACRES:
NA.21

€76.30


Estimated to ship on24 May 2025


Request a Bulk Order

Quality Level

form

liquid

greener alternative product characteristics

Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

29.5-34.5% sequestering agent content (complexometric)
4.3-5.3% Free alkali (as NH3)

pH

13 (20 °C in H2O)

density

1.294 g/cm3 at 20 °C

greener alternative category

storage temp.

15-25°C

General description

Extran® AP 17 is an alkaline laboratory cleaning agent. It cleans reliably and prevents residues from being carried over to the subsequent analysis, and ensuring accurate results. It is free from organic surfactants and emulsifiers but contains complexing agents. Therefore, it can be used in both soft and hard water.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is environmentally friendly having bio-degradable active ingredients. Click here for more information.

Application

Extran® AP 17 is an active universal cleaning agent for the main wash cycle which cleans and removes even heavily soiled items. Particularly suitable for the removal of starch and protein residues. Extran® AP 17 alkaline does not foam even during heavy agitation in a washing machine.

Linkage

Replaces: EX0991-4; EX0991

Analysis Note

sequestering agent content (complexometric): 29.5 - 34.5 %
Density (d 20 °C/ 4 °C): 1.27 - 1.31
Free alkali (as NH₃): 4.3 - 5.3 %

Legal Information

EXTRAN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service