Skip to Content
Merck
All Photos(1)

Key Documents

1.30103

Millipore

HY-LiTE® Hygiene Swab

Dry, sterile sampling swabs for use with HY-LiTE® ATP test kits and HY-LiTE® 2/2A luminometers.

Synonym(s):

Surface ATP swabs, Surface ATP samplers

Sign Into View Organizational & Contract Pricing

Select a Size

50 UNITS
€56.90

€56.90


Estimated to ship on02 June 2025


Request more information

Select a Size

Change View
50 UNITS
€56.90

About This Item

UNSPSC Code:
41112601
NACRES:
NA.74

€56.90


Estimated to ship on02 June 2025


Request more information

Quality Level

product line

HY-LiTE®

packaging

pkg of 50 (swabs)

manufacturer/tradename

HY-LiTE®

application(s)

food and beverages
surface monitoring

compatibility

for use with HY-LITE® luminometer

storage temp.

2-30°C

General description

The HY-LiTE® Hygiene Swab is designed for use with HY-LiTE® pens and HY-LiTE® 2/2A luminometers for hygiene monitoring of surfaces in food and beverages production. It is an easy-to-use, rapid and reliable test system to assess the quality of processing equipment, surfaces, water samples and other environments.

Application

The HY-LiTE® Hygiene Swab is designed for use with HY-LiTE® 2/2A luminometers for hygiene monitoring of processing equipment surfaces, water samples and other environments in food and beverages production

Linkage

SKU 1.30103.0002 replaced by 1301030001

Legal Information

HY-LITE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service