Skip to Content
Merck
All Photos(1)

Key Documents

1.25165

Millipore

Eshmuno® CPS Resin

MiniChrom Column Eshmuno® CPS, volume 5 mL, prepacked column

Synonym(s):

MiniChrom Column Eshmuno® CPS, 5ml

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
Inquire

This item is not able to be purchased as a guest user. For further information, please contact us at +43 1 3602 77 47 43or

.


Select a Size

Change View
1 EA
Inquire

About This Item

UNSPSC Code:
41115711

This item is not able to be purchased as a guest user. For further information, please contact us at +43 1 3602 77 47 43or

.

ligand

(Sulfoisobutyl)

material

polypropylene housing

product line

Eshmuno®

form

prepacked column

parameter

≤8 bar pressure
500 cm/hr max. flow rate

column I.D. × L

8 mm × 100 mm

volume

5 mL

matrix active group

Polyvinyl Ether

mean particle size

50 μm

capacity

~160 mL, resin binding capacity

application(s)

recombinant protein

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

8x100mm

Features and Benefits

Eshmuno® CPS chromatography resin enables:

  • Outstanding binding capacities at elevated conductivity levels (≥ 10 mS/cm)
  • Excellent separation efficiencies at high protein loadings
  • Ease of implementation and selection of process parameters through strong cation exchanger chemistry without hydrophobic groups
  • Established Fab fragments, recombinant human growth factor, and recombinant proteins purification protocols
  • High flow rate versus pressure-flow behavior
  • Excellent removal of impurities
  • Robust and safe packing procedures
  • Strong chemical stability
  • Compatibility with 1 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl and storage solutions 0.1 M NaOH

Packaging

Plastic bag(s)

Legal Information

Eshmuno is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service