Skip to Content
Merck
All Photos(1)

Key Documents

12-509

Sigma-Aldrich

Goat Anti-Mouse IgG (H+L) Antibody, Rhodamine conjugate

Upstate®, from goat

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€168.00

€168.00


Estimated to ship on21 May 2025



Select a Size

Change View
500 μG
€168.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

€168.00


Estimated to ship on21 May 2025


biological source

goat

Quality Level

conjugate

TRITC conjugate
rhodamine conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Upstate®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

shipped in

wet ice

target post-translational modification

unmodified

Application

Goat anti-Mouse IgG (H+L) Antibody, Rhodamine conjugate detects level of Mouse IgG (H+L) & has been published & validated for use in FC, IF, IC, IH.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Quality

Routinely evaluated by immunomicroscopy, flow cytometry or FACS analysis, as well as other antibody based fluorescent assays requiring lot-to-lot consistency

Physical form

0.02M potassium phosphatase, pH 7.2, 150mM NaCl, 0.01% sodium azide and 10mg/ml BSA

Storage and Stability

Lyophilized: Stable for 2 years from date of shipment at 4°C. Rehydrated: Stable for 1 year at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nuclear magnetic resonance structure revealed that the human polyomavirus JC virus agnoprotein contains an ?-helix encompassing the Leu/Ile/Phe-rich domain.
Coric, P; Saribas, AS; Abou-Gharbia, M; Childers, W; White, MK; Bouaziz, S; Safak, M
Journal of virology null
A Sami Saribas et al.
Journal of cellular physiology, 233(3), 2343-2359 (2017-07-20)
Agnoprotein (Agno) is an important regulatory protein of JC virus (JCV), BK virus (BKV) and simian virus 40 (SV40) and these viruses are unable to replicate efficiently in the absence of this protein. Recent 3D-NMR structural data revealed that Agno
Peter M Crapo et al.
Journal of biomedical materials research. Part A, 96(4), 673-681 (2011-01-27)
Prior studies have demonstrated that smooth muscle cell (SMC) proliferation, migration, and extracellular matrix production increase with hydrostatic pressure in vitro. We have engineered highly compliant small-diameter arterial constructs by culturing primary adult baboon arterial SMCs under pulsatile perfusion on
Saswat Mohapatra et al.
ACS omega, 6(11), 7815-7828 (2021-03-30)
Here, we demonstrate an interesting strategy of modulating mitochondrial reactive oxygen species (ROS) using the organic electron acceptor molecule carbonyl-bridged bithiazole attached with bis-trifluoroacetophenone (BBT). This molecule was found to affect complex I activity. It has the propensity to bind

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service