Skip to Content
Merck
All Photos(1)

Key Documents

12-507

Sigma-Aldrich

Goat Anti-Rabbit IgG (H+L) Antibody, FITC conjugate

Upstate®, from goat

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€207.00

€207.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
500 μG
€207.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

€207.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

goat

Quality Level

conjugate

FITC conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

rabbit

manufacturer/tradename

Upstate®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

shipped in

dry ice

target post-translational modification

unmodified

Application

Detect Rabbit IgG (H+L) using this Goat anti-Rabbit IgG (H+L) Antibody, FITC conjugate validated for use in FC, IF, IC, IH.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Quality

Routinely evaluated by immunomicroscopy, flow cytometry or FACS analysis, as well as other antibody based fluorescent assays requiring lot-to-lot consistency

Physical form

Lyophilized powder

Storage and Stability

Store for 1 year at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jana Subrova et al.
International journal of molecular sciences, 23(4) (2022-02-27)
Left ventricular hypertrophy (LVH) is a major risk factor for adverse cardiovascular events. Recently, a novel candidate gene encoding the carboxypeptidase X member 2 (CPXM2) was found to be associated with hypertension-induced LVH. CPXM2 belongs to the M14 family of
Ho-Chen Lin et al.
Toxicological sciences : an official journal of the Society of Toxicology, 158(1), 151-163 (2017-05-02)
Silver nanoparticles (AgNPs) enter the central nervous system through the blood-brain barrier (BBB). AgNP exposure can increase amyloid beta (Aβ) deposition in neuronal cells to potentially induce Alzheimer's disease (AD) progression. However, the mechanism through which AgNPs alter BBB permeability
Larisa V Antonova et al.
Frontiers in pharmacology, 7, 230-230 (2016-08-16)
The combination of a natural polymer poly(3-hydroxybutyrate-co-3-hydroxyvalerate) (PHBV) and a synthetic hydrophobic polymer poly(ε-caprolactone) (PCL) is promising for the preparation of biodegradable and biocompatible small-diameter vascular grafts for bypass surgery. However, physico-mechanical properties and endothelialization rate of PHBV/PCL grafts are
Shibin He et al.
International journal of molecular sciences, 21(19) (2020-10-01)
Histone acetylation plays an important role in regulation of chromatin structure and gene expression in terms of responding to abiotic stresses. Histone acetylation is modulated by histone deacetylases (HDACs) and histone acetyltransferases. Recently, the effectiveness of HDAC inhibitors (HDACis) for
Fei Gao et al.
Reproduction (Cambridge, England), 148(4), 377-387 (2014-07-18)
The Wilms' tumour 1 (WT1) gene originally identified as a tumour suppressor associated with WTs encodes a zinc finger-containing transcription factor that is expressed in multiple tissues and is an important regulator of cellular and organ growth, proliferation, development, migration

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service