Skip to Content
Merck
All Photos(1)

Key Documents

12-488

Sigma-Aldrich

Rabbit Anti-Mouse IgMµ Antibody, mu chain

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€204.00

€204.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
500 μG
€204.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

€204.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

shipped in

dry ice

target post-translational modification

unmodified

Application

Rabbit anti-Mouse IgMμ Antibody, mu chain is an antibody against Mouse IgMμ for use in IP & WB.

Quality

Suitable for use as a bridging antibody for detection of IgM monoclonal antibodies.

Physical form

0.02M potassium phosphate, 0.15M NaCl, pH 7.2, 0.01% (w/v) sodium azide

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bmi1 is essential for leukemic reprogramming of myeloid progenitor cells.
J Yuan,M Takeuchi,M Negishi,H Oguro,H Ichikawa,A Iwama
Leukemia null
Makiko Mochizuki-Kashio et al.
Blood, 118(25), 6553-6561 (2011-11-02)
Polycomb-group (PcG) proteins are essential regulators of hematopoietic stem cells (HSCs). In contrast to Bmi1, a component of Polycomb repressive complex 1 (PRC1), the role of PRC2 and its components in hematopoiesis remains elusive. Here we show that Ezh2, a
Pengsheng Chen et al.
Molecular & cellular proteomics : MCP, 18(7), 1320-1329 (2019-04-18)
BRCA1-associated protein 1 (BAP1) is a member of the ubiquitin C-terminal hydrolase family of deubiquitinating enzymes and is implicated in transcriptional regulation. The BAP1 gene is mutated in about 10% of patients with ccRCC, the most common form of renal
Hideyuki Oguro et al.
The Journal of experimental medicine, 209(3), 445-454 (2012-02-22)
Polycomb-group (PcG) proteins form the multiprotein polycomb repressive complexes (PRC) 1 and 2, and function as transcriptional repressors through histone modifications. They maintain the proliferative capacity of hematopoietic stem and progenitor cells by repressing the transcription of tumor suppressor genes
Josie X Zhou et al.
The Journal of clinical investigation, 123(11), 4849-4858 (2013-11-13)
Inadequate functional β cell mass underlies both type 1 and type 2 diabetes. β Cell growth and regeneration also decrease with age through mechanisms that are not fully understood. Age-dependent loss of enhancer of zeste homolog 2 (EZH2) prevents adult

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service