Skip to Content
Merck
All Photos(1)

Documents

12-242

Sigma-Aldrich

Glycogen Synthase Peptide-2

Ala21, negative control for GSK-3β

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
eCl@ss:
32160405
NACRES:
NA.42

product name

Glycogen Synthase Peptide-2 (Ala21), Glycogen Synthase Peptide-2 (Ala21) primarily used in Kinase Assays.

Quality Level

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

UniProt accession no.

shipped in

wet ice

Quality

Routinely evaluated for use as a negative control for assay of GSK-3β (Catalog # 14-306) activity in a phosphorylation assay. Not to be used as a substrate for GSK3.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S R Kimball et al.
The Journal of biological chemistry, 273(47), 30945-30953 (1998-11-13)
The present study was designed to investigate the mechanism through which leucine and histidine regulate translation initiation in L6 myoblasts. The results show that both amino acids stimulate initiation and coordinately regulate the activity of eukaryotic initiation factor eIF2B. The
M P Coghlan et al.
Chemistry & biology, 7(10), 793-803 (2000-10-18)
Glycogen synthase kinase-3 (GSK-3) is a serine/threonine protein kinase, the activity of which is inhibited by a variety of extracellular stimuli including insulin, growth factors, cell specification factors and cell adhesion. Consequently, inhibition of GSK-3 activity has been proposed to
Regulation and functions of the glycogen synthase kinase-3 subfamily.
Woodgett, J R
Seminars in cancer biology, 5, 269-275 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service