1.19682
PharmPrep® P Si 100, 20µm
PharmPrep® P Si100, 20µm. CAS 7631-86-9, EC Number 231-545-4, chemical formula SiO₂.
Synonym(s):
Silica, Silicon dioxide
Select a Size
€130.00
Estimated to ship on21 May 2025
Select a Size
About This Item
€130.00
Estimated to ship on21 May 2025
Recommended Products
material
spherical silica gel
Quality Level
potency
>5000 mg/kg LD50, skin (Rabbit)
parameter
≤25 bar max. pressure
pore size
10 nm mean pore size
pH
7 (20 °C, 100 g/L in H2O, slurry)
bp
2230 °C (lit.)
mp
>1600 °C (lit.)
bulk density
200‑800 kg/m3
application(s)
plasma purification
separation technique
reversed phase
storage temp.
2-30°C
SMILES string
O=[Si]=O
InChI
1S/O2Si/c1-3-2
InChI key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Analysis Note
Purity (Fe): ≤ 25 ppm
Purity (Na): ≤ 25 ppm
theoretical number of plates (2-Nitrotoluene): ≥ 7000 1 / m
Pressure: ≤ 10 bar
Pore volume: 0.8 - 0.9 ml/g
Specific surface area (BET): 320 - 400 m²/g
Particle Size (d10): 10.0 - 15.0 µm
Particle Size (d50): 16.0 - 23.0 µm
Particle Size (d90): 25.0 - 40.0 µm
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service