Skip to Content
Merck
All Photos(1)

Key Documents

119140

Sigma-Aldrich

NECA

≥99% (HPLC), solid, adenosine receptor agonist, Calbiochem®

Synonym(s):

Adenosine Receptor Agonist, NECA, (2S,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-N-ethyl-3,4-dihydroxytetrahydrofuran-2-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€109.00

€109.00


Estimated to ship on02 April 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€109.00

About This Item

Empirical Formula (Hill Notation):
C12H16N6O4
CAS Number:
Molecular Weight:
308.29
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€109.00


Estimated to ship on02 April 2025


Request a Bulk Order

Product Name

Adenosine Receptor Agonist, NECA,

Assay

≥99% (HPLC)

Quality Level

form

solid

potency

14 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mM

shipped in

wet ice

storage temp.

2-8°C

SMILES string

[n]2(c3ncnc(c3nc2)N)C1O[C@@H]([C@H]([C@H]1O)O)C(=O)NCC

InChI

1S/C12H16N6O4/c1-2-14-11(21)8-6(19)7(20)12(22-8)18-4-17-5-9(13)15-3-16-10(5)18/h3-4,6-8,12,19-20H,2H2,1H3,(H,14,21)(H2,13,15,16)/t6-,7+,8-,12?/m0/s1

InChI key

JADDQZYHOWSFJD-QQIVLSGASA-N

General description

A cell-permeable adenosine analog that acts a potent non-selective agonist of adenosine receptors (Ki = 14 nM, 20 nM, 2.4 µM and 6.2 nM for A1, A2A, A2B, A3, respectively). Increases intracellular cAMP production (EC50 = 3.1 µM in A2B expressing CHO cells). Shown to increase glucagon release in a dose-dependent manner and inhibit insulin release at low concentrations. Although at higher concentration some insulin release is observed. Also, displays a wide range adenosine-dependent effects, such as blocking platelet aggregation and inhibiting DNA synthesis. When administered at reperfusion, it is shown reduce infarction and block the formation of the mitochondrial permeability transition pore by activating p70S6 kinase.

Biochem/physiol Actions

Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Forster, K., et al. 2006. Basic Res. Cardiol.101, 319.
de Zwart, M., et al. 1998. Nucleosides Nucleotides.17, 969.
Phillis, J. et al. 1986, Pharm. Biochem. Behavior.24, 263
Durcan, M. J., et al. 1989. Pharmacol Biochem Behav.32, 487.
Bacher, S., et al. 1982. Naunyn Schmiedebergs Arch Pharmacol.329, 67.
Cusack, N., et al. 1981. Br J Pharm.72, 443.
Londos, C., et al. 1980. Proc Natl Acad Sci.77, 2551.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service