Skip to Content
Merck
All Photos(1)

Key Documents

119135

Sigma-Aldrich

Adenosine A1 Receptor Agonist I, CPA

Synonym(s):

Adenosine A1 Receptor Agonist I, CPA

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€63.90

€63.90


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€63.90

About This Item

Empirical Formula (Hill Notation):
C15H21N5O4
CAS Number:
Molecular Weight:
335.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€63.90


Estimated to ship on14 April 2025


Request a Bulk Order

Assay

≥99% (HPLC)

Quality Level

form

solid

potency

0.8 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

1 M HCl: 100 mM
DMSO: 100 mM
ethanol: 25 mM

shipped in

ambient

storage temp.

2-8°C

SMILES string

[n]2(c3ncnc(c3nc2)NC4CCCC4)C1O[C@@H]([C@H]([C@H]1O)O)CO

InChI

1S/C15H21N5O4/c21-5-9-11(22)12(23)15(24-9)20-7-18-10-13(16-6-17-14(10)20)19-8-3-1-2-4-8/h6-9,11-12,15,21-23H,1-5H2,(H,16,17,19)/t9-,11-,12-,15?/m1/s1

InChI key

SQMWSBKSHWARHU-FJFSNTMWSA-N

General description

A potent and selective agonist of adenosine A1 receptor (A1R) (Ki = 2.3 nM, 790 nM, 18.6 µM, 43nM for human A1, A2A, A2B, and A3, respectively).

Biochem/physiol Actions

Primary Target
A1
Reversible: yes

Warning

Toxicity: Standard Handling (A)

Preparation Note

slight warming is required for complete solubilization in ethanol

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Migita, H., et al. 2008. J Neurosci Res.86, 2820.
Burgdorf, C. et al. 2005. J Cardiovasc Pharmacol.45, 1.
Klotz, K. N., et al. 2000. Naunyn Schmiedebergs Arch Pharmacol. 362, 382.
Mironov, S., et al. 1990. J Neurophysiol.81, 1.
Monopoli, A., et al. 1994. Arzneimittelforschung.44, 1305.
Concas, A., et al. 1993. J Pharmacol Exp Ther.267, 844.
Lohse, M. J., et al. 1988. Nauryn Schmiedebergs Arch Pharmacol.337, 687.
Coffin, V. L., et al. 1987. J Pharmacol Exp Ther.241, 76.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service