Skip to Content
Merck
All Photos(1)

Key Documents

1.17182

Millipore

MAS-100 Iso NT® Power Supply

including cord EU, GB, US, JP

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€626.00

€626.00


Estimated to ship on28 May 2025


Request more information

Select a Size

Change View
1 PKG
€626.00

About This Item

UNSPSC Code:
41122409
NACRES:
NA.74

€626.00


Estimated to ship on28 May 2025


Request more information

material

plastic hardware

Agency

non-compendial

feature

electronic

packaging

pkg of 1 kit

application(s)

air monitoring
pharmaceutical

compatibility

for use with MAS-100 Iso NT®

storage temp.

no temp limit

General description

MAS-100 ISO NT® Power Supply is an additional accessory designed to be specifically used for MAS-100 ISO NT® air sampler. The MAS-100 ISO NT® air samplers has been specially developed for microbial monitoring of ambient air is isolators, RABS and high-grade cleanrooms. The Power supply has an input voltage of 100 to 240 V AC (47 to 63 Hz, 1.5 to 0.7 A) and output voltage of 24 V DC ±5% (2.7 A). The cable length is 1.5m on the AC side and 1.7m on the DC side with a Cable conductor cross-chapter of 18AWG.

Application

MAS-100 ISO NT® Power Supply is an additional accessory. MAS-100 ISO NT® air sampling devices are extensively used in Pharmaceutical applications.

Legal Information

MAS-100 ISO NT is a registered trademark of MBV AG

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service